1-Heptanesulfonic acid sodium salt monohydrate for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
41127-0250
41127-1000
AC41127-0050
AC41127-5000
TS41127-0250EA
208
USD
TS41127-0250
TS41127-1000
TS41127-0050
TS41127-5000
1-Heptanesulfonic acid sodium salt monohydrate for HPLC
1-Heptanesulphonic acid sodium salt monohydrate
Formula:
C₇H₁₅NaO₃S·1H₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149550 CAS Number: 207300-90-1 EINECS: 245-210-5 |
Specification Test Results
Appearance | White Powder or flakes |
Infrared spectrum | Conforms |
Loss on drying | 7.4 % to 9 % (100°C, 3 hrs) (vacuum) |
Solubility | (10 % w/v in water) Clear colorless |
UV | at 260 nm A: ≤0.01 |
Titration after Ion exchange | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...