di-Potassium hydrogen phosphate, anhydrous 99+% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

21547-0010 21547-2500 AC21547-0050
TS21547-0010EA 228 USD
TS21547-0010 TS21547-2500 TS21547-0050
di-Potassium hydrogen phosphate, anhydrous 99+% for analysis
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤1 % (130°C)
pH 8.5 to 9.6 (5 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR