Sinapic acid predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

11555-0250 11555-0010 11555-0050
TS11555-0250EA 402 USD
TS11555-0250 TS11555-0010 TS11555-0050
Sinapic acid predominantly trans 98%
Sinapic acid
Formula: C₁₁H₁₂O₅
MW: 224.21 g/mol
Boiling Pt: 378
Melting Pt: –99 °C (dec.)
Density: 0.873 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00004401
CAS Number: 530-59-6
EINECS: 203-745-1
UN: 1213
ADR: 3 II
Merck Index: 13,05147

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20°C, 589 nm)
Water ≤0.03 %
Density (20°C) 0.871 to 0.875 kg/l
GC ≥97.5 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR