Benzil 99+%

Supplier: Thermo Scientific Chemicals
Warning

10541-5000 10541-0025 10541-1000
TS10541-5000EA 88 USD
TS10541-5000 TS10541-0025 TS10541-1000
Benzil 99+%
Benzil
Formula: (C₆H₅)C(O)C(O)(C₆H₅)
Boiling Pt: 346…348 °C (760 mmHg)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00003080
CAS Number: 134-81-6
EINECS: 205-157-0
Merck Index: 13,01078

Order Now

Specification Test Results

Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 93 to 97 °C
GC ≥99.0 %
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤500 ppm
Potassium (K) ≤200 ppm
Sodium (Na) ≤500 ppm
Carbon compounds (as C) ≤0.01 %
Insoluble matter ≤0.02 % (in dilute HCl)
Nitrogen compounds (as N) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR