Cyanogen bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

11078-1000 11078-0250 11078-5000
TS11078-1000EA 159 USD
TS11078-1000 TS11078-0250 TS11078-5000
Cyanogen bromide 97%
Cyanogen bromide
Formula: CBrN
Boiling Pt: 61.4 °C (1013 hPa)
Melting Pt: 49…54 °C
Flash Pt: >65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1,I
Merck Index: 12,02763

Order Now

Specification Test Results

Appearance White Crystals
Titration Argentometric 96.0 to 103.0 %
Identification Bromides : positive
Clarity of solution (Aqueous) Passes test
Clarity of alcohol solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR