Lead(II) nitrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead dinitrate

42385-5000 42385-0025 42385-0050
TS42385-5000EA 106 USD
TS42385-5000 TS42385-0025 TS42385-0050
Lead(II) nitrate ACS
Lead (II) nitrate
Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 470 °C (dec.)
Density: 4.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 II
Merck Index: 13,05432

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤50 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR