Tetrabutylammonium tetrafluoroborate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35309-1000
AC35309-0050
AC35309-0250
TS35309-1000EA
287
USD
TS35309-1000
TS35309-0050
TS35309-0250
Tetrabutylammonium tetrafluoroborate 98%
Tetrabutylammonium tetrafluoroborate
Formula:
C₁₆H₃₆BF₄N Melting Pt: 156…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011634 CAS Number: 429-42-5 EINECS: 207-058-8 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...