Barium nitrate ACS

Supplier: Thermo Scientific Chemicals
Danger

19255-0025 AC19255-5000
TS19255-0025EA 239 USD
TS19255-0025 TS19255-5000
Barium nitrate ACS
Barium nitrate
Formula: Ba(NO₃)₂
Melting Pt: 592 °C
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
UN: 1446
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥99.0 %
pH 5.0 to 8.0 (5 % at 25.0°C)
Chloride (Cl) ≤5 ppm
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤2 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.1 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR