Sodium peroxide 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: di-Sodium peroxide

20770-1000 AC20770-0025 AC20770-0250 AC20770-5000
TS20770-1000EA 140 USD
TS20770-1000 TS20770-0025 TS20770-0250 TS20770-5000
Sodium peroxide 96%
Sodium peroxide
Formula: Na₂O₂
MW: 77.98 g/mol
Melting Pt: 460 °C (dec.)
Density: 2.8 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 13,08728

Order Now

Specification Test Results

Appearance Yellow Granules (May contain a small quantity of discolored material from the manufacturing process)
Titration with KMnO4 ≥95 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR