Hydrogen peroxide 30% (w/w) in water unstabilized ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
41188-0025
41188-5000
AC41188-0010
AC41188-1000
TS41188-0025EA
393
USD
TS41188-0025
TS41188-5000
TS41188-0010
TS41188-1000
Hydrogen peroxide 30% (w/w) in water unstabilized ACS
Hydrogen peroxide
Formula:
H₂O₂ Boiling Pt: ∼107 °C (1013 hPa) Melting Pt: –33 °C Density: 1.11 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1,II |
Specification Test Results
Appearance | Clear solution |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Titration with KMnO4 | 29.0 to 32.0 % |
Chloride (Cl) | ≤2 ppm |
Nitrate (NO3) | ≤2 ppm |
Phosphate (PO4) | ≤2 ppm |
Sulfate (SO4) | ≤5 ppm |
Ammonium (NH4) | ≤5 ppm |
Iron (Fe) | ≤0.5 ppm |
Heavy metals (as Pb) | ≤1 ppm |
Titrable acid | ≤0.0006 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...