Sodium nitrate 99+% for biochemistry
Supplier: Thermo Scientific Chemicals
|
Warning
|
44681-0010
TS44681-0010EA
108
USD
TS44681-0010
TS44681-2500
Sodium nitrate 99+% for biochemistry
Sodium nitrate
Formula:
NaNO₃ Boiling Pt: 380 °C (760 mmHg) Melting Pt: 306 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011119 CAS Number: 7631-99-4 UN: 1498 ADR: 5.1,III |
Specification Test Results
Appearance | White Crystalline powder |
pH | 5.5 to 8.3 |
Titration with HClO4 | ≥99.0 % |
Chloride (Cl) | ≤0.001 % |
Iodate (IO3) | ≤0.0005 % |
Nitrite (NO2) | ≤0.001 % |
Phosphate (PO4) | ≤0.0005 % |
Sulfate (SO4) | ≤0.003 % |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤0.0003 % |
Magnesium (Mg) | ≤0.002 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...