Sodium perchlorate, anhydrous ACS

Supplier: Thermo Scientific Chemicals
Danger

44742-0025 44742-5000 AC44742-1000
TS44742-0025EA 602 USD
TS44742-0025 TS44742-5000 TS44742-1000
Sodium perchlorate, anhydrous ACS
Sodium perchlorate
Formula: ClNaO₄
Melting Pt: 482 °C
Storage Temperature: Ambient
MDL Number: MFCD00003480
CAS Number: 7601-89-0
UN: 1502
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
Titration Argentimetric 98.0 to 102.0 %
pH 6.0 to 8.0 (5% solution at 25.0°C)
Chloride (Cl) ≤0.003 %
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤5 ppm
Potassium (K) ≤0.05 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR