Lithium carbonate 99.999% (trace metals basis), extra pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
19336-1000
AC19336-0100
TS19336-1000EA
222
USD
TS19336-1000
TS19336-0100
Lithium carbonate 99.999% (trace metals basis), extra pure
Lithium carbonate
Formula:
Li₂CO₃ MW: 73.89 g/mol Melting Pt: 720 Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 13,05549 |
Specification Test Results
Appearance | White Powder |
Total trace metal impurities | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...