Sodium borohydride 99%, granules, VenPure™ SF, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
44850-1000
AC44850-0025
AC44850-0100
AC44850-5000
TS44850-1000EA
88
USD
TS44850-1000
TS44850-0025
TS44850-0100
TS44850-5000
Sodium borohydride 99%, granules, VenPure™ SF, pure
Sodium borohydride
Formula:
NaBH₄ MW: 37.83 g/mol Melting Pt: 400 (dec.) Flash Pt: 69 Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 UN: 1426 ADR: 4.3 I Merck Index: 13,08664 |
Specification Test Results
Appearance | White Granules |
Assay | ≥98.5 % (Hydrogen evolution) |
Iron (Fe) | ≤5 ppm |
Silicon dioxide (SiO2) | 21 to 500 ppm |
Fines | ≤8.5 % (< 0.5 cm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...