Sodium borohydride 98%, granules ∼ 3 mm

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

41947-1000 AC41947-5000
TS41947-1000EA 86 USD
TS41947-1000 TS41947-5000
Sodium borohydride 98%, granules ∼ 3 mm
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance White Granules
Titration Iodometric ≥97.5 %
Water ≤0.2 % (Coulometric)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR