Cyclohexene 99% stabilised, pure

Supplier: Thermo Scientific Chemicals
Danger

15484-0010 154840100 154845000
TS15484-0010EA 111 USD
TS15484-0010 TS154840100 TS154845000
Cyclohexene 99% stabilised, pure
Cyclohexene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 83
Melting Pt: -104
Density: 0.81 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001539
CAS Number: 110-83-8
EINECS: 203-807-8
UN: 2256
ADR: 3 II
Merck Index: 13,02756

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4480 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 100 ppm 2-tert-Butyl-4-methylphenol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR