Methylcyclohexane 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

12658-0025 126580010 126580050 126580100
TS12658-0025EA 268 USD
TS12658-0025 TS126580010 TS126580050 TS126580100
Methylcyclohexane 99%, Extra Pure
Methylcyclohexane
Formula: C₆H₁₁CH₃
MW: 98.19 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –126.3 °C
Density: 0.77 g/cm³
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 13,06074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4240 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA
Residue after evaporation ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR