Cyclopentane 97%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pentamethylene
11148-0025
111480010
111481000
TS11148-0025EA
518
USD
TS11148-0025
TS111480010
TS111481000
Cyclopentane 97%, extra pure
Cyclopentane
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 49 Melting Pt: -93.3 Density: 0.75 g/cm³ Flash Pt: –7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001356 CAS Number: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3 II Merck Index: 13,02769 |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4050 to 1.4070 (20°C, 589 nm) |
GC | ≥96.0 % |
Residue after evaporation | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...