Hexafluorobenzene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perfluorobenzene

12054-1000 12054-0100 12054-5000
TS12054-1000EA 328 USD
TS12054-1000 TS12054-0100 TS12054-5000
Hexafluorobenzene 99%
Hexafluorobenzene
Formula: C₆F₆
MW: 186.06 g/mol
Boiling Pt: 81…82 °C (743 mmHg)
Melting Pt: 3.7…4.1 °C
Density: 1.612 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000288
CAS Number: 392-56-3
EINECS: 206-876-2
UN: 1993
ADR: 3,II
Merck Index: 13,04705

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3760 to 1.3780 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR