Glycerine (synthetic) 99+%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

33203-0010 33203-0025 332030100 332031000
TS33203-0010EA 93 USD
TS33203-0010 TS33203-0025 TS332030100 TS332031000
Glycerine (synthetic) 99+%, pure
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.261 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4750 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR