Ethylene glycol, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane

44423-0050 44423-0010
TS44423-0050EA 115 USD
TS44423-0050 TS44423-0010
Ethylene glycol, Technical Grade
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (760 mmHg)
Melting Pt: -13
Density: 1.113 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR