Trichloroacetic acid (TCA) 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
TCA, Trichloroethanoic acid, Aceto-Caustin, TCA
15213-0010
15213-0025
15213-5000
152130100
TS15213-0010EA
186
USD
TS15213-0010
TS15213-0025
TS15213-5000
TS152130100
Trichloroacetic acid (TCA) 99%, Extra Pure
Trichloroacetic acid (TCA)
Formula:
Cl₃CCOOH MW: 163.39 g/mol Boiling Pt: 196 °C (760 mmHg) Melting Pt: 52…58 °C Density: 1.62 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00004177 CAS Number: 76-03-9 EINECS: 200-927-2 UN: 1839 ADR: 8,II Merck Index: 13,09700 |
Specification Test Results
Appearance | White Crystalline powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 52 to 58 °C |
Water | ≤0.5 % (K.F.) |
Titration with NaOH | ≥98.5 % |
Iron (Fe) | ≤10 ppm |
Sulfuric acid | ≤0.3 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...