Chloroform ≥99.8% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
Stabilised with ethanol
42355-0025
42355-0040
42355-0250
42355-5000
423550010
423550251
TS42355-0025EA
175
USD
TS42355-0025
TS42355-0040
TS42355-0250
TS42355-5000
TS423550010
TS423550251
Chloroform ≥99.8% stabilized ACS, meets analytical specifications of Reagent Ph. Eur.
Chloroform
Formula:
CHCl₃ MW: 119.38 g/mol Boiling Pt: 61 °C (760 mmHg) Melting Pt: –63 °C Density: 1.48 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00000826 CAS Number: 67-66-3 UN: 1888 ADR: 6.1,III |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Boiling point | 60 to 62 °C |
Density | (20/4 °C) 1.474 to 1.483 |
GC | ≥99.80 % (Excluding stabilizer) |
Color scale | ≤10 APHA |
Stabilizer | 0.5 to 1.0 % Ethanol |
Residue after evaporation | ≤0.001 % |
Substance darkened by H2SO4 | Passes test |
Free chlorine | (Cl) Passes test |
Lead (Pb) | ≤0.05 ppm |
Acid and chloride | Passes test |
Acetone and aldehyde | Passes test |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...