Acetonitrile 99+%, extra pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
14952-0050
149520010
149520025
149520250
TS14952-0050EA
459
USD
TS14952-0050
TS149520010
TS149520025
TS149520250
Acetonitrile 99+%, extra pure
Acetonitrile
Formula:
H₃CCN Boiling Pt: 81…82 °C (760 mmHg) Melting Pt: –46 °C Density: 0.781 g/cm³ Flash Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.3 % (K.F.) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Residue after evaporation | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...