Strontium chloride hexahydrate 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

31508-5000 AC31508-1000
TS31508-5000EA 200 USD
TS31508-5000 TS31508-1000
Strontium chloride hexahydrate 99+% ACS
Strontium chloride hexahydrate
Formula: SrCl₂·6H₂O
MW: 266.62 g/mol
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00149865
CAS Number: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric 99.0 to 103.0 %
pH 5.0 to 7.0 (5 % at 25°C)
Sulfate (SO4) ≤10 ppm
Barium (Ba) ≤500 ppm
Calcium (Ca) ≤500 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR