Methanol-[D₄] 99.6+ atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

32536-0100 32536-0500
TS32536-0100EA 153 USD
TS32536-0100 TS32536-0500
Methanol-[D₄] 99.6+ atom % D for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3
Melting Pt: -99
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.60 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR