Zinc bromide 99.999% (trace metals basis), extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) bromide

AC21277-0025 AC21277-0100 AC21277-0500
TS21277-0025EA 38 USD
TS21277-0025 TS21277-0100 TS21277-0500
Zinc bromide 99.999% (trace metals basis), extra pure
Zinc bromide
Formula: Br₂Zn
Boiling Pt: 670…680 °C (1013 hPa)
Melting Pt: 394 °C
Density: 4.22 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 13,10182

Order Now

Specification Test Results

Appearance White Crystalline powder
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR