1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (S,S)-Et-DUPHOS-Rh, (+)-1,2-BIS((2S,5S)-2,5-DIETHYLPHOSPHOLANO)BENZENE(CYCLOOCTADIENE)RHODIUM(L)TETRAFLUOROBORATE, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

AC43519-1000 AC43519-2500
TS43519-1000EA 129 USD
TS43519-1000 TS43519-2500
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Formula: C₃₀H₄₆BF₄P₂Rh
Storage Temperature: Ambient
MDL Number: MFCD01862465
CAS Number: 213343-64-7

Order Now

Specification Test Results

Appearance Orange to red to brown Powder
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 52.4 to 56.8 % C
Specific optical rotation -40° to -50° (20°C, 589 nm) (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR