Silver sulfadiazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: sulfadiazine Silver, Sulphadiazine Silver

AC45965-0050 AC45965-0250
TS45965-0050EA 65 USD
TS45965-0050 TS45965-0250
Silver sulfadiazine 98%
Silver sulfadiazine
Formula: C₁₀H₉AgN₄O₂S
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00072101
CAS Number: 22199-08-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR