Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40,8% Rh)

Supplier: Thermo Scientific Chemicals

Synonyms: (1,5-Cyclooctadiene)rhodium chloride dimer

AC21130-0050 AC21130-5000
TS21130-0050EA 4326 USD
TS21130-0050 TS21130-5000
Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40,8% Rh)
Chloro(1,5-cyclooctadiene)rhodium (I) dimer
Formula: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
Melting Pt: 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012415
CAS Number: 12092-47-6

Order Now

Specification Test Results

Appearance Orange to orange-brown Crystalline powder or crystals
Infrared spectrum Conforms
Rhodium (Rh) ≥40.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR