Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt

AC41050-0250 AC41050-1000 AC41050-5000
TS41050-0250EA 33 USD
TS41050-0250 TS41050-1000 TS41050-5000
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Potassium xanthogenate (Potassium O-ethyl dithiocarbonate)
Formula: C₃H₅KOS₂
Melting Pt: 209…214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004931
CAS Number: 140-89-6
EINECS: 205-439-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Titration with HClO4 ≥97.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR