Palladium(II) trifluoroacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

AC31702-0010 AC31702-0050
TS31702-0010EA 111 USD
TS31702-0010 TS31702-0050
Palladium(II) trifluoroacetate 97%
Palladium (II) trifluoroacetate
Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
Melting Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now

Specification Test Results

Appearance Tan to brown Powder or crystals
Infrared spectrum Conforms
Assay 30.9 to 33.2 % (Pd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR