4-Bromopyridinium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

AC10742-0050 AC10742-0250
TS10742-0050EA 110 USD
TS10742-0050 TS10742-0250
4-Bromopyridinium chloride 98%
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
Melting Pt: 313…315 °C
Storage Temperature: Ambient
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4

Order Now

Specification Test Results

Appearance White to light yellow-pink Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
UV at 400 nm A: ≤0.004
Color scale ≤10 APHA
Stabilizer Approx. 1 % w/w Ethanol
Peroxides ≤5 ppm (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR