2-Bromethylamine hydrobromide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

AC10693-0010 AC10693-0025 AC10693-0250 AC10693-1000 AC10693-2500 AC10693-5000
TS10693-0010EA 806 USD
TS10693-0010 TS10693-0025 TS10693-0250 TS10693-1000 TS10693-2500 TS10693-5000
2-Bromethylamine hydrobromide 99%
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR