Ammonium hydrogen sulfate, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium bisulfate, Ammonium bisulphate

AC44591-0010 AC44591-0025
TS44591-0010EA 85 USD
TS44591-0010 TS44591-0025
Ammonium hydrogen sulfate, extra pure
Ammonium hydrogen sulphate
Formula: H₅NO₄S
MW: 115.11 g/mol
Boiling Pt: 350
Melting Pt: 138.85 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051073
CAS Number: 7803-63-6
UN: 2506
ADR: 8 II
Merck Index: 12,00526

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with NaOH ≥97.5 %
Chloride (Cl) ≤0.005 %
Phosphate (PO4) ≤0.003 %
Insoluble matter ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR