Barium titanate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

AC19686-0025 AC19686-5000
TS19686-0025EA 214 USD
TS19686-0025 TS19686-5000
Barium titanate 99%
Barium titanate
Formula: BaO₃Ti
Melting Pt: 1625 °C
Density: 6.08 g/cm³
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
Merck Index: 12,01029

Order Now

Specification Test Results

Appearance Off-white Powder
Assay 0.988 to 1.000 (BaO + SrO)/TiO2 Mole Ratio
Particle size 0.85 to 1.45 µm (Horiba, D50)
Silicon dioxide (SiO2) ≤0.02 %
Impurity ≤0.15 % (SrO)
Loss on ignition ≤0.5 %
Aluminium oxide (Al2O3) ≤0.04 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR