Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ruthenium trichloride
AC19721-0020
AC19721-0100
AC19721-0500
TS19721-0020EA
124
USD
TS19721-0020
TS19721-0100
TS19721-0500
Ruthenium(III) chloride, anhydrous 99.9% (trace metals basis)
Ruthenium (III) chloride
Formula:
Cl₃Ru Melting Pt: >500 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011208 CAS Number: 10049-08-8 EINECS: 233-167-5 UN: 3260 ADR: 8,II Merck Index: 12,08455 |
Specification Test Results
Appearance | Black Powder or crystals |
Assay | 48 to 50 % (Ru) |
Total trace metal impurities | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...