Aluminium sulfate octadecahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trade termforalum, Aluminum sulfate octadecahydrate, Aluminum(III) sulfate octadecahydrate, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate

AC40059-0025 AC40059-5000
TS40059-0025EA 439 USD
TS40059-0025 TS40059-5000
Aluminium sulfate octadecahydrate 98+% ACS
Aluminium sulphate octadecahydrate
Formula: Al₂(SO₄)₃·18H₂O
MW: 666.43 g/mol
Melting Pt: 86 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR