Triethylsilane 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC44616-0010 AC44616-1000
TS44616-0010EA 895 USD
TS44616-0010 TS44616-1000
Triethylsilane 99%, AcroSeal™
Triethylsilane
Formula: C₆H₁₆Si
Boiling Pt: 107…108 °C (760 mmHg)
Density: 0.72 g/cm³
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00009018
CAS Number: 617-86-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR