Titanium(IV) oxide 98.0-100.5%

Supplier: Thermo Scientific Chemicals

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

AC27737-0010 AC27737-0100
TS27737-0010EA 82 USD
TS27737-0010 TS27737-0100
Titanium(IV) oxide 98.0-100.5%
Titanium (IV) oxide
Formula: TiO₂
Boiling Pt: 2900 °C (760 mmHg)
Melting Pt: 1855 °C
Density: 4.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Specification Test Results

Appearance White to almost white Powder
Acidity/alkalinity Passes test
Titration 98.0 to 100.5 %
Antimony (Sb) ≤100 ppm
Arsenic (As) ≤5 ppm
Barium (Ba) passes test
Iron (Fe) ≤200 ppm
Soluble matter ≤0.5 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR