Nickel(II) nitrate hexahydrate 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC22315-0025 AC22315-0100 AC22315-5000
TS22315-0025EA 442 USD
TS22315-0025 TS22315-0100 TS22315-5000
Nickel(II) nitrate hexahydrate 99% for analysis
Nickel (II) nitrate hexahydrate
Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C (760 mmHg)
Melting Pt: 56.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1,III
Merck Index: 12,06596

Order Now

Specification Test Results

Appearance Green to blue-green Crystals and/or chunks
Titration Complexometric ≥98.5 %
Chloride (Cl) ≤50 ppm
Aluminium (Al) ≤10 ppm
Arsenic (As) ≤5 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤20 ppm
Chromium (Cr) ≤5 ppm
Cobalt (Co) ≤500 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤20 ppm
Manganese (Mn) ≤20 ppm
Sulfur (S) ≤100 ppm
Zinc (Zn) ≤10 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR