Copper(II) bromide, anhydrous 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric bromide, Cuprous bromide

AC20633-0025 AC20633-0050 AC20633-1000 AC20633-5000
TS20633-0025EA 985 USD
TS20633-0025 TS20633-0050 TS20633-1000 TS20633-5000
Copper(II) bromide, anhydrous 99+%, extra pure
Copper (II) bromide
Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900
Melting Pt: 498
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8 III
Merck Index: 13,02656

Order Now

Specification Test Results

Appearance Dark gray to gray-black Crystalline powder
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤5000 ppm
Sulfate (SO4) ≤2000 ppm
Insoluble matter ≤0.05 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR