Chlorotrimethylsilane 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMCS, TMSC, Trimethylchlorosilane

AC42643-1000
TS42643-1000EA 89 USD
TS42643-1000
Chlorotrimethylsilane 98%, AcroSeal™
Chlorotrimethylsilane
Formula: C₃H₉ClSi
Boiling Pt: 57 °C (760 mmHg)
Melting Pt: –58 °C
Density: 0.85 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000502
CAS Number: 75-77-4
UN: 1298
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3890 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR