Tin (II) sulfate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC22366-0025
AC22366-1000
AC22366-5000
TS22366-0025EA
502
USD
TS22366-0025
TS22366-1000
TS22366-5000
Tin (II) sulfate 95%
Tin (II) sulphate
Formula:
O₄SSn Melting Pt: 360 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011246 CAS Number: 7488-55-3 EINECS: 231-302-2 Merck Index: 12,08947 |
Specification Test Results
Appearance | White to off-white Powder or crystals and/or lumps |
Titration Iodimetric | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...