Potassium sulfate 99+% for biochemistry

Supplier: Thermo Scientific Chemicals

AC42421-0010 AC42421-2500
TS42421-0010EA 183 USD
TS42421-0010 TS42421-2500
Potassium sulfate 99+% for biochemistry
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Appearance White Crystals
Solubility (10 g in 150 ml water) Clear colorless
pH 5.5 to 8.5 (5 % in water)
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR