Zinc sulfide 99.99% (trace metals basis) < 10 µm

Supplier: Thermo Scientific Chemicals

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

AC22365-0250 AC22365-1000 AC22365-5000
TS22365-0250EA 39 USD
TS22365-0250 TS22365-1000 TS22365-5000
Zinc sulfide 99.99% (trace metals basis) < 10 µm
Zinc sulphide
Formula: SZn
MW: 97.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now

Specification Test Results

Appearance White to light yellow Powder
Total trace metal impurities ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR