Barium hydroxide octahydrate 99.95%

Supplier: Thermo Scientific Chemicals
Danger

AC37782-0050 AC37782-0250
TS37782-0050EA 50 USD
TS37782-0050 TS37782-0250
Barium hydroxide octahydrate 99.95%
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or flakes and/or chunks
Total trace metal impurities ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR