Vinyltrimethoxysilane 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethoxy(vinyl)silane

AC43306-0010 AC43306-1000
TS43306-0010EA 215 USD
TS43306-0010 TS43306-1000
Vinyltrimethoxysilane 98%, AcroSeal™
Vinyltrimethoxysilane
Formula: C₅H₁₂O₃Si
MW: 148.23 g/mol
Boiling Pt: 123 °C (760 mmHg)
Melting Pt: 0 °C
Density: 0.968 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00008605
CAS Number: 2768-02-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3950 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR