Vinyltrimethoxysilane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trimethoxy(vinyl)silane
AC21652-1000
AC21652-5000
TS21652-1000EA
46
USD
TS21652-1000
TS21652-5000
Vinyltrimethoxysilane 98%
Vinyltrimethoxysilane
Formula:
C₅H₁₂O₃Si MW: 148.23 g/mol Boiling Pt: 123 °C (760 mmHg) Melting Pt: 0 °C Density: 0.968 g/cm³ Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008605 CAS Number: 2768-02-7 EINECS: 220-449-8 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3915 to 1.3935 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...