Zinc acetate dihydrate 98%, extra pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate
AC20764-0010
AC20764-0050
AC20764-5000
TS20764-0010EA
98
USD
TS20764-0010
TS20764-0050
TS20764-5000
Zinc acetate dihydrate 98%, extra pure
Zinc acetate dihydrate
Formula:
(H₃CCOO)₂Zn·2H₂O Melting Pt: 237 °C Density: 1.84 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00066961 CAS Number: 5970-45-6 EINECS: 209-170-2 UN: 3077 ADR: 9,III Merck Index: 12,10256 |
Specification Test Results
Appearance | Colorless to white Powder or crystals |
Titration Complexometric | ≥97.5 % |
Iron (Fe) | ≤100 ppm |
Lead (Pb) | ≤50 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...